Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINA7 Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA127075

Catalog No. PIPA127075


Add to cart

Description

Description

PA1-27075 detects Thyroxine Binding Globulin from human samples. PA1-27075 will not react with related serum proteins. PA1-27075 has been successfully used in radioimmune assay procedures. The PA1-27075 immunogen is Thyroxine Binding Globulin purified from human serum (> 95% pure by SDS-PAGE).

Specifications

Specifications

SERPINA7
Polyclonal
Unconjugated
SERPINA7
Liquid
Sheep
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6906
Radioimmune Assays
4.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05543
SERPINA7
Thyroxine Binding Globulin purified from human serum (> 95% pure by SDS-PAGE).
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit