Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-cell lymphoma invasion and metastasis 2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant TIAM2.

Manufacturer:  Abnova Corporation H00026230A01

Catalog No. 89-004-585


Add to cart

Description

Description

This gene encodes a guanine nucleotide exchange factor. A highly similar mouse protein specifically activates ras-related C3 botulinum substrate 1, converting this Rho-like guanosine triphosphatase (GTPase) from a guanosine diphosphate-bound inactive state to a guanosine triphosphate-bound active state. The encoded protein may play a role in neural cell development. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: DVHPEAEQQPGPESGEGQKGGEQPKLVRGHFCPIKRKANSTKRDRGTLLKAQIRHQSLDSQSENATIDLNSVLEREFSVQSLTSVVSEECFYETESHG
Specifications

Specifications

T-cell lymphoma invasion and metastasis 2
Unconjugated
50% glycerol
NM_001010927
FLJ41865|STEF
Mouse
50 uL
Membrane Receptors
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant TIAM2.
TIAM2
Serum
TIAM2
TIAM2 (NP_001010927, 527 a.a. ~ 624 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
26230
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit