Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TIFA, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192507

Catalog No. NBP192507

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TIFA Polyclonal antibody specifically detects TIFA in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TIFA
0.05mg/mL
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500
Affinity Purified
TIFA
The immunogen recognized by this antibody maps to the antigen sequence: FEDADTEETVTCLQMTVYHPGQLQCGIFQSISFNREKLPSSEVVKFGRNSNICHYTFQDKQVSRVQFSLQLFKKFNSSVLSFEI
Immunogen affinity purified
RUO
Primary
92610
Human
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Putative MAPK-activating protein PM14, Putative NF-kappa-B-activating protein 20, T2BPMGC20791, T6BP, TIFAA, TRAF2 binding protein, TRAF2-binding protein, TRAF6 binding protein, TRAF-interacting protein with a forkhead-associated domain, TRAF-interacting protein with FHA domain-containing protein A, TRAF-interacting protein with forkhead-associated domain
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only