Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIFA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526591

Catalog No. PIPA526591

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Adapter protein which mediates the IRAK1 and TRAF6 interaction following IL-1 stimulation, resulting in the downstream activation of NF-kappa-B and AP-1 pathways. Induces the oligomerization and polyubiquitination of TRAF6, which leads to the activation of TAK1 and IKK through a proteasome-independent mechanism.
Specifications

Specifications

TIFA
Polyclonal
PBS with 0.09% sodium azide
Q96CG3
T2BP, T6BP, TIFAA, TRAF-interacting protein with FHA domain-containing protein A, TRAF-interacting protein with a forkhead-associated domain, TRAF2 binding protein, TRAF2-binding protein, TRAF6 binding protein, putative MAPK-activating protein PM14, putative NF-kappa-B-activating
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
92610
Western Blot
Unconjugated
TIFA
Liquid
TIFA
KLH conjugated synthetic peptide between 52-79 amino acids from the N-terminal region of human TIFA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit