Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TIGAR/C12orf5, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149534AF488

Catalog No. NBP149534X


Add to cart

Description

Description

TIGAR/C12orf5 Polyclonal antibody specifically detects TIGAR/C12orf5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TIGAR/C12orf5
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
TIGAR
A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Q9NQ88
chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
Rabbit
IgG
0.1 ml
Lipid and Metabolism
Primary
57103
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only