Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-1/KIM-1/HAVCR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$204.00 - $482.50
Specifications
Antigen | TIM-1/KIM-1/HAVCR |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056421T
|
Novus Biologicals
NB10056421SS |
0.025 mg |
Each for $204.00
|
|
|||||
NB10056421
|
Novus Biologicals
NB10056421 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TIM-1/KIM-1/HAVCR | |
Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
26762 | |
A synthetic peptide (SPLYSYTTDGNDTVT) of human TIM-1 protein was used as immunogen for this antibody. This sequence corresponds to amino acids (aa) 248-262 of NP_036338 and aa 84-98 of EAW61615.1. Isoforms of TIM-1 have been described. For example, NP_036338 is 364 aa and EAW61615.1 is 194 aa. EAW61615.1 is referred to as isoform CRA_a. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
HAVCR1 | |
IgG | |
Immunogen affinity purified | |
Isoforms of different molecular weights have been described for TIM-1. The sequence used for immunogen is conserved between TIM-1 (364aa, NP_036338) and TIM-1 isoform CRA_a (194aa, EAW61615.1). Therefore, the molecular weight observed in western blots may vary depending on the forms present. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title