Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD365 (TIM-1) Mouse anti-Human, PE, Clone: 1D12, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564820

Catalog No. BDB564820


Add to cart

Description

Description

The 1D12 monoclonal antibody specifically binds to CD365, the T-cell immunoglobulin mucin receptor 1 (TIM-1). TIM-1 is expressed on kidney epithelial cells, T cells, and some hematopoietic and non-hematopoietic cells. CD365 (TIM-1) is a type 1 transmembrane glycoprotein that serves as a receptor for hepatitis A virus and is encoded by the HAVCR1 (Hepatitis A virus cellular receptor 1) gene. TIM-1 also serves as a receptor for phosphatidylserine which is exposed on the surface of apoptotic cells. TIM-1 can reportedly mediate the uptake of apoptotic cells through the recognition of phosphatidylserine and thus help maintain tissue homeostasis and self-tolerance. TIM-1 is likewise known as Kidney injury molecule 1 (KIM-1). It is highly expressed by cancerous kidneys, and upregulated in the proximal tubular epithelium and shed into the urine during acute and chronic kidney injury. CD365 (TIM-1) also functions as a costimulatory molecule for immune cells. It is expressed by activated CD4+ T cells and regulates the effector functions (eg, enhanced cytokine production) and survival of differentiated T cells, including those mediating Th2-like immune responses. Other ligands have been described for TIM-1 including TIM-4 and LMIR5 (also known as CD300b) which are expressed by myeloid cells. With respect to disease associations, the HAVCR1 gene has been linked to asthma, allergy, and some autoimmune diseases.

Specifications

Specifications

CD365
1D12
TIM1; TIM; TIMD1; TIMD-1; HAVCR1; HAVCR-1; HAVCR; KIM1; KIM-1
Affinity Purified
Human TIM-1 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit