Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-1 Mouse anti-Human, PE, Clone: 219211, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523614

Catalog No. PIMA523614


Add to cart

Description

Description

This antibody does not cross-react with recombit mouse (rm) TIM-1, rmTIM-2, or rhTIM-3.

The human form of TIM-1 was initially discovered as a membrane glycoprotein through which the hepatitis A virus can gain entry into a cell. It was also identified as kidney injury molecule 1 (Kim-1), a predicted adhesion molecule that is upregulated on the surfaces of kidney epithelia. It is also expressed on T helper 2 (Th2) cells of the immune system, and following the binding of its natural ligand TIM-4, stimulates T cell expansion and cytokine production in response to viral challenge. It has been suggested that hyperactivation of TIM-1 leads to an increased level of Th2 responsiveness and asthma susceptibility, and antibodies to TIM-1 may therefore be a novel approach to treating asthma.
Specifications

Specifications

TIM-1
Monoclonal
PE
HAVCR1
Liquid
HAVCR1
Mouse myeloma cell line NS0-derived recombit human TIM-1/KIM-1/HAVCR Ser21-Thr288
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
219211
Buffered saline solution with BSA and 0.09% sodium azide
Q96D42
HAVCR, HAVCR-1, KIM-1, KIM1, TIM-1, TIM1, TIMD1
Mouse
IgG2B
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
26762
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit