Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-3 Mouse anti-Human, Clone: 183551, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen TIMP-3
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625
Clone 183551
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB973SP
View Documents
R&D Systems
MAB973SP
25 ug
Each for $125.00
Add to cart
 
MAB973100
View Documents
R&D Systems
MAB973100
100 ug
Each for $356.00
Add to cart
 
MAB973500
View Documents
R&D Systems
MAB973500
500 ug
Each for $839.00
Add to cart
 
Description

Description

TIMP-3 Monoclonal antibody MAB973 Specifically detects TIMP-3 in Human samples. It is validated for ELISA Capture, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

TIMP-3
183551
Unconjugated
P35625
Monoclonal
Primary
Detects human TIMP-3 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human TIMP-1, -2, or -4 is observed.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625
Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA
Purified
Mouse
IgG1
7078
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit