Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121146

Catalog No. PA121146

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The tissue inhibitors of metalloproteinases (TIMPs) are naturally occurring proteins that specifically inhibit matrix metalloproteinases and regulate extracellular matrix turnover and tissue remodeling by forming tightly bound inhibitory complexes with the MMPs. Thus, TIMPs maintain the balance between matrix destruction and formation. An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors.

PA1-21146 detects TIMP-3 in rabbit, human and baboon samples. This antibody will not detect rat, bovine, avian, or porcine samples.PA1-21146 has been successfully used in immunohistochemistry (paraffin) procedures.The PA1-21146 immunogen is a synthetic peptidecorresponding to the C-terminal portion of human TIMP-3.
Specifications

Specifications

TIMP3
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P35625
HSMRK222, K222, K222TA2, SFD
Rabbit
IgG
7mL
Store at 4°C, do not freeze
Primary
7078
Immunohistochemistry (Paraffin)
Unconjugated
TIMP3
Purified
TIMP3
Synthetic peptidecorresponding to the C-terminal portion of human TIMP-3.
Purified
RUO
Antibody
Polyclonal
Human, Non-human Primate, Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit