Learn More
TIMP3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526133
Catalog No. PIPA526133
Add to cart
Description
Description
This antibody is predicted to react with bovine, non-human primate, rat and rabbit based on sequence homology.
TIMP3 is one of inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix (ECM).
Specifications
Specifications
TIMP3
Polyclonal
Unconjugated
TIMP3
Liquid
TIMP3
KLH conjugated synthetic peptide between 113-139 amino acids from the central region of human TIMP3
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
P35625, P39876
HSMRK222, K222, K222TA2, SFD, MIG-5 protein, TIMP-3, metalloproteinase inhibitor 3, protein MIG-5, tissue inhibitor of metalloproteinases 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
21859, 7078