Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TIMP4 Clone: 13H4L13, Invitrogen™

TIMP4 Monoclonal antibody 701317 from Thermo Scientific™ Pierce™ specifically detects TIMP4 in Human samples, and it is validated for WB, IF, ICC, FACS and ELISA

Manufacturer:  Invitrogen 701317

Encompass

Catalog No. 70-131-7


Add to cart

Description

Description

Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.

701317 detects TIMP4 in Human samples. 701317 has been successfully used in WB, IF, ICC, FACS and ELISA procedures. The immunogen for 701317 is proprietary.

Applications:

ELISA, FACS, Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

TIMP4
Unconjugated
Q99727
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
13H4L13
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
proprietary
Protein A
Primary
7079
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit