Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TIN2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP154837

Catalog No. NBP154837

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TIN2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TINF2
Synthetic peptides corresponding to TIN2. The peptide sequence was selected from the middle region of TIN2. Peptide sequence SDEEENGQGEGKESLENYQKTKFDTLIPTLCEYLPPSGHGAIPVSSCDCR.
50 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Rat: 85%.
Human
Western Blot
Western Blot 1:100-1:2000
Q9BSI4
(TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
26277
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only