Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TINAGL1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523896

Catalog No. PIPA523896

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TINAGL1
Polyclonal
PBS with 0.09% sodium azide
Q9GZM7, Q99JR5
ARG1, LCN7, LIECG3, TINAGRP, OLRG-2, P3ECSL, TIN Ag-related protein, TIN-Ag-RP, TINAG-like 1, androgen-regulated gene 1, glucocorticoid-inducible protein 5, lipocalin 7, oxidized-LDL responsive gene 2, tubulointerstitial nephritis antigen-like, tubulointerstitial nephritis antigen-related prot
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64129, 94242
Western Blot
Unconjugated
TINAGL1
Liquid
TINAGL1
KLH conjugated synthetic peptide between 370-398 amino acids from the C-terminal region of human TINAGL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit