Learn More
TIP5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen 491037
Catalog No. 49-103-7
Add to cart
Description
Description
TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
Specifications
Specifications
TIP5
Polyclonal
whole serum with <0.1% sodium azide
Q9UIF9, Q91YE5
Bromodomain adjacent to zinc finger domain protein 2A, Transcription termination factor I-interacting protein 5, TTF-I-interacting protein 5, Baz2a, Kiaa0314, Tip5
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
ChIP assay, Western Blot
Unconjugated
BAZ2A
Liquid
BAZ2A
Recombit protein TIP5
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11176, 116848