Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIRAP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112816

Catalog No. 01-675-808


Add to cart

Description

Description

A suggested positive control for this product is liver, kidney, spleen, skeletal muscle or heart lysate.

Toll-like receptors (TLRs) are evolutionarily conserved patternrecognition molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The signaling of these TLRs is kept under tight control by the expression of endogenous inhibiting proteins such as RP105, a recently identified homolog of TLR4. This protein, in association with MD-1, interacts with and inhibits the TLR4/MD-2 signaling pathway.. While MD-2 can directly bind to LPS, one of the activating molecules of TLR4, the function of MD -1 is less well-known. It has been suggested however, that the RP105/MD-1 complex influences antibody production mediated by both TLR4/MD-2 and TLR2 receptor complexes.
Specifications

Specifications

TIRAP
Polyclonal
PBS with 0.02% sodium azide
P58753
Mal, wyatt
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
114609
Western Blot
Unconjugated
TIRAP
Liquid
TIRAP
Synthetic peptide corresponding to residues near the middle of human TIRAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit