Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Rabbit anti-Human, Mouse, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128063

Catalog No. PA128063


Add to cart

Description

Description

PA1-28063 detects TLE1 from mouse, human, Xenopus laevis and rat samples. Expected to cross-react with chicken (92% identity with immunogen) due to sequence homology. PA1-28063 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), and Western blot applications. By Western Blot a band of ~83.5 kDa is detected. The PA1-28063 immunogen is Synthetic peptide: DDKKHHDAEHHRDREPGT conjugated to BSA, corresponding to amino acids 181-198 of Human TLE1. Store at 4°C short term. For extended storage, store at -20°C, avoiding freeze/thaw cycles.

Groucho (TLE1) (Transduction-like enhancer protein 1, ESG1) is a member of the Groucho family of transcriptional corepressors and is involved in oligomerization, association with chromatin and interaction with histone H3.
Specifications

Specifications

TLE1
Polyclonal
Unconjugated
TLE1
Liquid
TLE1, tle1.S
synthetic peptide DDKKHHDAEHHRDREPGT conjugated to BSA, corresponding to amino acids 181-198 of Human TLE1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat, Xenopus laevis
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% ammonium sulfate and no preservative; pH 7.2
Q04724, Q62440
RP11-203D19.1, ESG, ESG1, GRG1
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
21885, 362533, 7088, 779361
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit