Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515374

Catalog No. PIPA515374

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The Tousled-like kinases, first described in Arabadopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.[supplied by OMIM]
Specifications

Specifications

TLK1
Polyclonal
PBS with 0.09% sodium azide
Q9UKI8
Serine/threonine-protein kinase tousled-like 1, Tousled-like kinase 1, PKU-beta
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9874
Immunohistochemistry, Western Blot
Unconjugated
TLK1
Liquid
TLK1
KLH conjugated synthetic peptide between 720-750 amino acids from the C-terminal region of human TLK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit