Learn More
TLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA517576
Catalog No. PIPA517576
Add to cart
Description
Description
It is not recommended to aliquot this antibody.
The Tousled-like kinases, first described in Arabidopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.
Specifications
Specifications
TLK1
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q9UKI8, Q8C0V0
SNARE protein kinase SNAK; serine threonine protein kinase; serine/threonine-protein kinase tousled-like 1
Rabbit
IgG
100 μL
-20°C
Primary
228012, 311118, 9874
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
TLK1
Liquid
TLK1
Synthetic peptide corresponding to residues adjacent to Ser183 of human TLK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat