Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBPL1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA528088

Catalog No. PIPA528088

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

PA5-28088 targets TLP in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-28088 immunogen is a synthetic peptide corresponding to a region within amino acids 123 and 186 of TLP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBPL1
Polyclonal
Unconjugated
P62380, P62340
TBPL1
Synthetic peptide corresponding to a region within amino acids 123 and 186 of TLP
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
TBPL1
MGC:8389, MGC:9620, STUD, TLF, TLP, TRF2, 21-kDA TBP-like protein, TATA box-binding protein-like protein 1, TATA box-binding protein-related factor 2, TBP-related factor 2, TBP-related protein, second TBP of unique DNA protein
Rabbit
Antigen affinity chromatography
RUO
237336, 9519
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit