Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TLR1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177244

Catalog No. NBP177244

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TLR1
1 mg/mL
Western Blot 1- 4 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/mL, Immunohistochemistry-Paraffin 10 ug/ml, Immunofluorescence 20 ug/ml
CAG38593
CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Ion Exchange Chromatography
TLR1
Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human TLR1. The immunogen is located within amino acids 160 - 210 of TLR1.
86 kDa
0.1 mg
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary
7096
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only