Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146478

Catalog No. PIPA146478


Add to cart

Description

Description

The target sequence has 90% sequence homology with bovine. Suggested positive control: antigen standard for TLR10 (transient overexpression lysate), Ramos whole cell extracts.

Toll-like receptors (TLRs) are membrane proteins on many cells involved with innate immunity such as macrophages, dendritic cells, neutrophils, mucosal epithelial cells, and endothelial cells. Ten mammalian TLRs have been identified and all have distinct specificities for microbial ligands. Upon recognition of a microbial product, TLRs initiate a signaling pathway to activate innate immunity.
Specifications

Specifications

TLR10
Polyclonal
Unconjugated
TLR10
Liquid
TLR10
Synthetic peptide corresponding to residues 1-100 of an N-terminal portion of human TLR10. [Swiss-ProtNo. Q9BXR5]
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
citric acid with 0.1% sodium azide; pH 7-8
Q9BXR5
Toll-like Receptor 10
Rabbit
IgG
100 μL
4° C, do not freeze
Primary
81793
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit