Learn More
TLR11 Rabbit anti-Mouse, Rat, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523103
Catalog No. PIPA523103
Add to cart
Description
Description
The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). TLR11 does not respond to known TLR ligands but instead respond specifically to uropathogenic bacteria. TLR11-deficient mice are highly susceptible to infection of the kidneys by uropathogenic bacteria, indicating that TLR11 may play a protective role in preventing infection of internal organs of the urogenital system.
Specifications
Specifications
TLR11
Polyclonal
FITC
Tlr11
Liquid
TLR11
Synthetic peptide (amino acids 911-926) of TLR11 protein.
Protein G
RUO
Antibody
Polyclonal
Mouse, Rat
Flow Cytometry
0.75 mg/mL
50mM sodium borate with 0.05% sodium azide
Q6R5P0
Gm287
Rabbit
IgG
100 μg
4° C, store in dark
Primary
239081, 498491