Learn More
TLR12 Rabbit anti-Mouse, Rat, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523100
Catalog No. PIPA523100
Add to cart
Description
Description
The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2).  TLR12 has been cloned that displays a distinct pattern of expression in macrophages and liver, kidney, and bladder epithelial cells. TLR12 does not respond to known TLR ligands.
Specifications
Specifications
TLR12
Polyclonal
FITC
Tlr12
Liquid
Rabbit
IgG
100 μg
4° C, store in dark
Primary
362604, 384059
Flow Cytometry
0.85 mg/mL
50mM sodium borate with 0.05% sodium azide
Q6QNU9
TLR12
Synthetic peptide (aa 743-756) of TLR12 protein.
Protein G
RUO
Antibody
Polyclonal
Mouse, Rat