Learn More
TLR2 Rabbit anti-Mouse, Human, Chimpanzee, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061119
Catalog No. 06-111-9MI
Add to cart
Description
Description
Anti-TLR2 Antibody is an antibody against TLR2 for use in western blot, immunohistochemistry (paraffin).
Specifications
Specifications
TLR2
Unconjugated
Affinity Purified
Rabbit
Affinity Purified
2-8°C for 12 months
Polyclonal
Chimpanzee, Human, Murine, Rhesus
Immunohistochemistry (Paraffin), Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
TLR2; TIL4; CD282
KLH-conjugated linear peptide corresponding to the extracellular domain of human TLR2.
100μg
Primary
NP_003255