Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523105

Catalog No. PIPA523105


Add to cart

Description

Description

Suggested positive control: mouse spleen lysate or protein.

The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1) and could constitute an important and unrecognized component of innate immunity in humans. The TRL family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). TLRs characterized so far activate the MyD88/interleukin-1 receptor-associated kinase (IRAK) signaling pathway. Ten human homologs of TLRs (TLR1-10) have been described (3). mTLR3 cDNA codes for a protein with approximate molecular weight of 100 kDa . TLR3 has a restricted expression pattern being expressed in dendritic cells (DC) (4-5). TLR3 mRNA expression was detected by in situ hybridization in DC and lymph nodes. The expression of TLR3 in a single cell type may indicate a specific role for this molecule in a restricted setting.
Specifications

Specifications

TLR3
Polyclonal
Unconjugated
TLR3
Liquid
TLR3
A mix of synthetic peptides corresponding to amino acids 135-150, 828-844, and 876-891 of TLR3/CD283 protein.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Porcine
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
O15455, Q99MB1
Toll-like Receptor 3, IIAE2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100037937, 142980, 7098
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit