Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523510

Catalog No. PIPA523510


Add to cart

Description

Description

Also classified as CD283, appears to be one of the major sensors of RNA viruses.  Viral recognition by endosomal TLRs induces Type I Interferon production and release.  TLR3 is the only human TLR which does not utilize MyD88 as an adaptor molecule instead utilizing TRIF in a signaling path which results in downstream activation of IRF-3 and NF-kB. TLR3 activation leads not only to Type I interferon induction but also to other inflammatory cytokines which result in DC maturation.   Through its recognition of dsRNA, TLR3 also appears to potentiate immune responses and TLR3 agonists have been exploited as vaccine adjuvants.
Specifications

Specifications

TLR3
Polyclonal
Unconjugated
TLR3
Liquid
TLR3
Synthetic peptide (356 KYLEYLNMDDNNIPSTKS 373) of TLR3 protein.
Protein G
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O15455
Toll-like Receptor 3, IIAE2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7098
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit