Learn More
anti-TLR4, APC, Clone: 610015, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems FAB6248A
Catalog No. FAB6248A
Add to cart
This item is not returnable. View return policy
Description
Description
TLR4 Monoclonal specifically detects TLR4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TLR4
610015
Flow Cytometry 10 uL/10^6 cells
O00206
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human TLR4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TLR1, 2, 3, 5, 7, 8, 9, 10, recombinant mouse (rm) TLR4, rmTLR6, rhTIRAP, rhIL-1 RI, rhIL-1 RII, rhIL-1 RAcP, rhST2, rhIL-18 R, rhIL-1 Rrp2, rhIL-18 R beta , rhSIGIRR, rhIL-1 RAPL1, rhIL-1 RAPL2, rhMD-1, rhMD-2, or rmRP105 is observed.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
TLR4
Mouse myeloma cell line NS0-derived recombinant human TLR4 Glu24-Lys631 Accession # O00206
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7099
Human