Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128112

Catalog No. PA128112

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Toll-like receptors (TLR) are higher conserved throughout evolution and have been implicated in the innate defense to many pathogens. In Drosphilia toll is required for the anti-fungal response, while the related 18-wheeler is involved in antibacterial defenses. In mammals, TLR identified as type 1 transmembrane signaling receptors with pattern recognition capabilities have been implicated in the innate host defense to pathogens.

PA1-28112 detects TLR4 from mouse, human samples.PA1-28112 has been successfully used in Western blot applications. Western blot detects a band of ~74-80kDa (predicted molecular weight 96kDa).The PA1-28112 immunogen is synthetic peptide: ELQVLDLSRCEIQTIEDG, corresponding to amino acids 39-56 of Mouse TLR4.Postive Control of partial recombinant mouse TLR4 (extracellular portion plus His-tag) suggested.Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications

Specifications

TLR4/CD284
Polyclonal
Unconjugated
TLR4
Purified
TLR4
synthetic peptide ELQVLDLSRCEIQTIEDG, corresponding to amino acids 39-56 of Mouse TLR4.
Protein G
RUO
Antibody
Polyclonal
Human, Murine
Immunohistochemistry, Western Blot
0.5mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O00206, Q9QUK6
Toll-like Receptor 4, ARMD10, CD284, TLR-4, TOLL, hToll, homolog of Drosophila toll
Rabbit
IgG
50μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
7099
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit