Learn More
TLR5 (CD285) Mouse anti-Human, Alexa Fluor 647, Clone: 624915, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564344
Catalog No. BDB564344
Add to cart
Description
Description
The 624915 monoclonal antibody specifically binds to TLR5 (Toll-like Receptor 5) which is also known as CD285, MELIOS, SLEB1, and TIL3. TLR5 is a type I transmembrane glycoprotein of the Toll-like Receptor family. It is expressed on mucosal epithelial cells and leucocytes, including monocytes and some immature dendritic cells and CD4-positive T cells. CD285 binds to bacterial flagellin. Flagellin binding induces TLR5 dimerization and intracellular signaling through MyD88 resulting in the cellular production of proinflammatory cytokines and immune system activation.
Specifications
Specifications
TLR5 (CD285)
624915
Toll-like receptor 5; MELIOS; SLEB1; TIL3
Affinity Purified
Human TLR5 Recombinant Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Human