Learn More
TLR6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523113
Catalog No. PIPA523113
Add to cart
Description
Description
TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.
Specifications
Specifications
TLR6
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q9Y2C9
Toll-like Receptor 6, CD antigen 286
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
TLR6
Liquid
TLR6
Synthetic peptide (aa 25-39, RIQFSDGNEFAVDKS) of TLR6/CD286 protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10333