Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLS Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611384

 View more versions of this product

Catalog No. BDB611384


Add to cart

Description

Description

Chromosomal translocations in leukemias and malignancies of non-hematopoietic tissues usually result in the fusion of genes and the production of novel fusion proteins. Translocated in liposarcoma (TLS/FUS) was identified through its fusion to the transcription factor CHOP in human myxoid liposarcoma. In addition, the fusion of TLS to the transcription factor ERG is involved in human acute myeloid leukemia. TLS contains an N-terminal Gln-, Ser-, and Tyr-rich region (QSY) that is thought to be a potent transactivator when fused with transcription factors. In its C-terminus, TLS contains a ribonucleoprotein consensus sequence (RNP-CS) and Arg-Gly-Gly (RGG) repeats that have been implicated in RNA binding. This feature allows it to function in shuttling of RNA from the nucleus. TLS also interacts with Ser-Arg proteins that regulate RNA splicing. Interestingly, TLS also interacts with the DNA binding domains of the estrogen, thyroid hormone, and glucocorticoid receptors. Thus, TLS is a multifunctional protein that is involved in RNA transport, nuclear receptor function, and gene transactivation when fused with transcription factors.

Immunofluorescence, Western Blotting

Specifications

Specifications

TLS
15
Unconjugated
Affinity Purified
Human TLS aa. 2-117
50μg
Cell Biology
Primary
Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.