Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM165 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525423

Catalog No. PIPA525423

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of TM165 remains unknown.
Specifications

Specifications

TMEM165
Polyclonal
Unconjugated
TMEM165
Liquid
TMEM165
KLH conjugated synthetic peptide between 17-45 amino acids from the N-terminal region of human TM165
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9HC07
TMPT27, TPARL, TPA regulated locus, transmembrane protein PT27, transmembrane protein TPARL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55858
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit