Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM2D3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525283

Catalog No. PIPA525283


Add to cart

Description

Description

The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. Several alternatively spliced transcript variants of this gene are described but the full length nature of some variants has not been determined. Multiple polyadenylation sites have been found in this gene.
Specifications

Specifications

TM2D3
Polyclonal
PBS with 0.09% sodium azide
Q9BRN9
BLP2, BBP-like protein 2, TM2 domain-containing protein 3, beta-amyloid-binding protein-like protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80213
Western Blot
Unconjugated
TM2D3
Liquid
TM2D3
KLH conjugated synthetic peptide between 22-50 amino acids from the N-terminal region of human TM2D3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit