Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF18 Mouse anti-Human, Clone: 211116, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523931

Catalog No. PIMA523931


Add to cart

Description

Description

This antibody stains human TM4SF18-transfected cells but not parent cells. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

TM4SF18
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q96CE8
AAB5, L6D
Mouse
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
116441
Flow Cytometry
211116
Unconjugated
TM4SF18
Lyophilized
TM4SF18
NS0 mouse myeloma cell line transfected with human TM4SF18 Met1-Ile201
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit