Learn More
TM7SF2 Rabbit anti-Human, Bovine, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061026
Catalog No. 06-102-6MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-TM7SF2 Antibody is a Rabbit Polyclonal Antibody for detection of TM7SF2 also known as Delta(14)-sterol reductase, Putative sterol reductase SR-1 and has been validated in western blot, immunocytochemistry.
Specifications
Specifications
TM7SF2
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Bovine, Human, Rhesus
Immunocytochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
TM7SF2; ANG1; NET47
KLH-conjugated linear peptide corresponding to human TM7SF2.
100μg
Primary
NP_003264.2