Learn More
TMC2 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00227W
Catalog No. PIOSR00227W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
May be required for the normal function of cochlear hair cells. Expressed at low, constant levels in temporal bone from embryonic day 14 to day 1 after birth. Increases by 8 to 16-fold at day 5, 10 and 20.
Specifications
Specifications
TMC2
Polyclonal
whole serum with no preservative
Q8R4P4
Beethoven, transmembrane channel-like protein 2, transmembrane cochlear-expressed protein 2, dn, Bth, CWEA1, 4933416G09Rik
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Tmc2
Lyophilized
TMC2
A synthetic peptide from the 1t cytoplasmic loop of mouse transmembrane channel-like protein 2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
192140