Learn More
TMC3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00153W
Catalog No. PIOST00153W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 µL of sterile water.
Specifications
Specifications
TMC3
Polyclonal
whole serum with no preservative
Q7Z5M5
TMC3
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TMC3
Lyophilized
TMC3
A synthetic from the 1t cytoplasmic loop of human transmembrane channel like protein 3
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
342125