Learn More
TMC5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00149W
Catalog No. PIOST00149W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability.
Specifications
Specifications
TMC5
Polyclonal
whole serum with no preservative
Q6UXY8
Transmembrane channel-like protein 5, TMC5
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TMC5
Lyophilized
TMC5
A synthetic peptide from the 1t extracellular loop of human transmembrane channel-like protein 5.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
79838