Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMC5 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific OST00158W

Catalog No. PIOST00158W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 100 uL of sterile water.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMC5
Polyclonal
Tmc5
TMC5
Rabbit
100 μL
Primary
Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q5M7W4
TMC5
A synthetic peptide from the 1t extracellular loop of rat transmembrane channel-like protein 5
RUO
365360
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit