Learn More
TMC6 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00223W
Catalog No. PIOSR00223W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Widely expressed in the Endoplasmic reticulum membrane; Multi-pass membrane protein.
Specifications
Specifications
TMC6
Polyclonal
whole serum with no preservative
Q7TN60
Transmembrane channel-like protein 6
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Tmc6
Lyophilized
TMC6
A synthetic peptide from the 6th lumenal mouse transmembrane channel-like protein 6
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
217353