Learn More
EVER2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00152W
Catalog No. PIOST00152W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability.
Epidermodysplasia verruciformis (EV) is an autosomal recessive dermatosis characterized by abnormal susceptibility to human papillomaviruses (HPVs) and a high rate of progression to squamous cell carcinoma on sun-exposed skin. EV is caused by mutations in either of two adjacent genes located on chromosome 17q25.3. Both of these genes encode integral membrane proteins that localize to the endoplasmic reticulum and are predicted to form transmembrane channels. This gene encodes a transmembrane channel-like protein with 8 predicted transmembrane domains and 3 leucine zipper motifs.
Specifications
Specifications
EVER2
Polyclonal
whole serum with no preservative
Q8IU68
TMC8, Epidermodysplasia verruciformis protein 2, EVER2, EVIN2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TMC8
Lyophilized
TMC8
A synthetic peptide from the 5th cytoplasmic loop of human transmembrane channel-like protein 8
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
147138