Learn More
EVER2 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00157W
Catalog No. PIOST00157W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 µL of sterile water.
Specifications
Specifications
EVER2
Polyclonal
whole serum with no preservative
Q7TN58
TMC8, Epidermodysplasia verruciformis protein 2, EVER2, EVIN2
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Tmc8
Lyophilized
TMC8
A synthetic peptide from the 1t cytoplasmic loop of mouse transmembrane channel-like protein 8
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
217356