Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMCO7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524500

Catalog No. PIPA524500

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TMCO7
Polyclonal
PBS with 0.09% sodium azide
Q9C0B7
transmembrane and coiled-coil domain-containing protein 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79613
Immunohistochemistry, Western Blot
Unconjugated
TANGO6
Liquid
TANGO6
KLH conjugated synthetic peptide between 904-933 amino acids from the C-terminal region of human TMCO7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit