Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM108 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526603

Catalog No. PIPA526603

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

TMEM108
Polyclonal
PBS with 0.09% sodium azide
Q6UXF1, Q8BHE4
CT124, cancer/testis antigen 124
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
66000, 81907
Western Blot
Unconjugated
TMEM108
Liquid
TMEM108
KLH conjugated synthetic peptide between 524-553 amino acids from the C-terminal region of human TMEM108
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit