Learn More
TMEM126A Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061037
Catalog No. 06-103-7MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-TMEM126A Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry, immunohistochemistry to detect TMEM126A also known as transmembrane protein 126A.
Specifications
Specifications
TMEM126A
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunocytochemistry, Immunohistochemistry, Western Blot
0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide
TMEM126A
KLH-conjugated linear peptide corresponding to human TMEM126A.
100μg
Primary
NP_115649