Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TMEM158, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191300

Catalog No. NBP191300


Add to cart

Description

Description

TMEM158 Polyclonal antibody specifically detects TMEM158 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TMEM158
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TMEM158
Synthetic peptide directed towards the middle region of human TMEM158. Peptide sequence AHGRAFFAAAFHRVGPPLLIEHLGLAAGGAQQDLRLCVGCGWVRGRRTGR.
Immunogen affinity purified
RUO
Primary
25907
Human
Western Blot
Western Blot 1:1000
NP_056259
BBP, transmembrane protein 158, transmembrane protein 158 (gene/pseudogene)
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only