Learn More
TMEM214 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523576
Catalog No. PIPA523576
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Transmembrane protein 214 (TMEM214) is a 77kD membrane protein which is widely expressed at high level. The gene encoding this protein maps to chromosome 2, at 2p23.3 TMEM214 interacts with LSM1, an SM-like protein, forming a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing. The detailed function of TMEM214 is still unknown, but a recent study showed that TMEM214 may be used to explore the contribution of human host factors of some infectious diseases such as Dengue fever, West Nile fever and yellow fever. Multiple isoforms of TMEM214 are known to exist.
Specifications
Specifications
TMEM214
Polyclonal
Unconjugated
TMEM214
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54867
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide
Q6NUQ4
TMEM214
KLH conjugated synthetic peptide between 38-66 amino acids from the N-terminal region of human TMEM214
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human