Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM26 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523477

Catalog No. PIPA523477


Add to cart

Description

Description

Transmembrane protein 26, or Tmem26, is a protein which spans the entire biological membrane and thus maintains three dimensional structures. Transmembrane proteins aggregate and precipitate in water. They require detergents or nonpolar solvents for extraction; although some of them (beta-barrels) can be also extracted using denaturing agents.
Specifications

Specifications

Tmem26
Polyclonal
Unconjugated
TMEM26
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
219623
Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
Q6ZUK4
TMEM26
Synthetic peptide (328 CRAQTSESGPSQRDWQNES 346) of Tmem26 protein.
Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit