Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TMEM48, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191604

Catalog No. NBP191604


Add to cart

Description

Description

TMEM48 Polyclonal antibody specifically detects TMEM48 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TMEM48
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
NDC1
Synthetic peptide directed towards the middle region of human TMEM48. Peptide sequence PPIIKYLALQDLMLLSQYSPSRRQEVFSLSQPGGHPHNWTAISRECLNLL.
Immunogen affinity purified
RUO
Primary
55706
Human
Western Blot
Western Blot 1:1000
NP_060557
FLJ10407, FLJ34120, NDC1, NET3, transmembrane protein 48
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only